Orto-perjodsyra ≥99.5%, AnalaR NORMAPUR® analytiskt reagens

Leverantör: VWR Chemicals
Fara

Synonyms: Perjodsyra

20593.151EA 1950 SEK
20593.151 20593.180
Orto-perjodsyra ≥99.5%, AnalaR NORMAPUR® analytiskt reagens
Orto-perjodsyra
Formel: H₅IO₆
MW: 227.94 g/mol
Smältpunkt: 122
Densitet: 1.3875 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011344
CAS nummer: 10450-60-9
EINECS: 233-937-0
UN: 3085
ADR: 5.1 (8) II
Merck Index: 13,07248
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39e1882cdf619581f467ea2429265a700b5205d5&vid=27e789fa0f30c08d62d5a8d0afc3acd8f6469c12&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Assay Min. 99.5 %
Insolubility in water Max. 50 ppm
Ignition residue (SO4) Max. 0.2 %
Cl + ClO3 + Br + BrO3 (as Cl) Max. 0.02 %
SO4 (Sulphate) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR