Perklorsyra 60%, AnalaR NORMAPUR® analytiskt reagens

Leverantör: VWR Chemicals
Fara

20583.240EA 1925 SEK
20583.240 20583.260 20583.327
Perklorsyra 60%, AnalaR NORMAPUR® analytiskt reagens
Perklorsyra
Formel: HClO₄
MW: 100.46 g/mol
Kokpunkt: 160…181 °C (1013 hPa)
Smältpunkt: –112,2 °C
Densitet: 1.53…1.768 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011325
CAS nummer: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1 (8) I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Assay 60 - 62 %
Free chlorine Max. 5 ppm
Ignition residue (SO4) Max. 30 ppm
Total N (Nitrogen) Max. 10 ppm
Cl (Chloride) Max. 3 ppm
ClO3 (Chlorate) Max. 10 ppm
PO4 + SiO4 (as SiO2) Max. 5 ppm
SO4 (Sulphate) Max. 10 ppm
Ag (Silver) Max. 5 ppm
As (Arsenic) Max. 0.05 ppm
Cu (Copper) Max. 0.1 ppm
Fe (Iron) Max. 1 ppm
Mg (Magnesium) Max. 10 ppm
Mn (Manganese) Max. 0.5 ppm
Pb (Lead) Max. 0.1 ppm

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR