Oxalsyra dihydrat ≥99%, GPR RECTAPUR®
Leverantör: VWR Chemicals
Varning
Synonyms: Ethanedioic acid dihydrate
20558.296EA 1050 SEK
20558.296 20558.365 20558.410 20558.460
Oxalsyra dihydrat ≥99%, GPR RECTAPUR®
Oxalsyra dihydrat
Formel: HO₂CCO₂H·2H₂O
MW: 126.07 g/mol
Kokpunkt: 149…160 °C (1013 hPa)
Smältpunkt: 101
Densitet: 1.65 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149102
CAS nummer: 6153-56-6
EINECS: 205-634-3
UN: 3261
ADR: 8 III
REACH: 01-2119534576-33
Merck Index: 13,06980
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Assay Min. 99 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 0.05 %
Fe (Iron) Max. 20 ppm
SPECIFIKATIONER
Environmentally Preferable