Salpetersyra 60%, AnalaR NORMAPUR® analytiskt reagens

Leverantör: VWR Chemicals
Fara

Synonyms: Nitric acid solution

20423.291EA 665 SEK
20423.291 20423.360
Salpetersyra 60%, AnalaR NORMAPUR® analytiskt reagens
Salpetersyra
Formel: HNO₃
MW: 63.01 g/mol
Kokpunkt: 87
Smältpunkt: -42
Densitet: 1.51
Storage Temperature: Omgivande
MDL Number: MFCD00011349
CAS nummer: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610

Order Now

Specifikation testresultat

Assay Min. 60.0 %
Density (20/4) 1.365 - 1.375
Heavy metals (as Pb) Max. 0.2 ppm
Ignition residue (SO4) Max. 10 ppm
Cl (Chloride) Max. 0.5 ppm
PO4 (Phosphate) Max. 2 ppm
SO4 (Sulphate) Max. 2 ppm
As (Arsenic) Max. 0.01 ppm
Fe (Iron) Max. 1 ppm
Mn (Manganese) Max. 0.4 ppm
Pb (Lead) Max. 0.05 ppm

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR