4-Aminophenylmercury acetate ≥90%, Calbiochem®, Sigma-Aldrich®

Leverantör: Merck Millipore Calbiochem
Fara

164610-700
164610-700EA 2300 SEK
164610-700
4-Aminophenylmercury acetate ≥90%, Calbiochem®, Sigma-Aldrich®
4-Aminophenylmercury acetate

An organomercurial agent used for the activation of latent MMPs in vitro. Acts by facilitating the loss of the enzyme propeptide domain through an autolytic cleavage known as the cysteine switch.

Formel: C₈H₉HgNO₂
Storage Temperature: Omgivande
CAS nummer: 6283-24-5
EINECS: 228-497-1
UN: 2025
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR