Methylparaben, Reference Standard
Leverantör: USP
1432005
1432005.EA 3950 SEK
1432005.
Methylparaben, Reference Standard
Standarder Organic Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifikat: US Pharmacopea Reference Standard
Order Now
SPECIFIKATIONER
Benämning Methylparaben
Environmentally Preferable
CAS-nr 99-76-3
Formel HOC₆H4CO2CH₃
Smältpunkt 125…128 °C (lit)
Förvaringstemperatur Ambient