Kaliumpermanganat 99,0 ACS

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: Permanganic acid potassium salt

14307.22 14307.A3
14307.22EA 605 SEK
14307.22 14307.A3
Kaliumpermanganat 99,0 ACS
Kaliumpermanganat

Purity: 99.0% min CAS No.: 7722-64-7 Molecular Formula: KMnO4 Formula Weight: 158.03 Physical Form: Crystalline Density: 2.7 Appearance: Dark purple Melting Point: 240° dec. Solubility: Partially soluble in water. Decomposes in organic solvents and acids Merck Reference: 14,7655 UN No.: 1490 MDL No.: MFCD00011364


In photography, tanning leather, purifying water, manufacturing mineral water, as an analytical lab reagent

Formel: KMnO₄
MW: 158.03 g/mol
Smältpunkt: 240 °C (dec.)
Densitet: 2.7
Storage Temperature: Omgivande
MDL Number: MFCD00011364
CAS nummer: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1 II
Merck Index: 14,7655

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR