Litium standard dissolved in oil, 1.000 µg/g Li in mineral oil Specpure® for wear metal analysis

Leverantör: Thermo Fisher Scientific

14215.18
14215.18EA 2175 SEK
14215.18
Litium standard dissolved in oil, 1.000 µg/g Li in mineral oil Specpure® for wear metal analysis
Standarder Cationic Standards Lithium Standards

Physical form: liquid
MDL No.: MFCD00134051

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR