Silver standard dissolved in oil, 1.000 µg/g Ag in mineral oil Specpure® for wear metal analysis

Leverantör: Thermo Fisher Scientific

14199.18
14199.18EA 2500 SEK
14199.18
Silver standard dissolved in oil, 1.000 µg/g Ag in mineral oil Specpure® for wear metal analysis
Standarder Cationic Standards Silver Standards

Physical form: liquid
MDL No.: MFCD00003397

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR