Washroom cleaners, SURE®
Leverantör: Diversey
100951914 100951912
141-1583EA 545 SEK
141-1583 141-1582
Washroom cleaners, SURE®
Rengöringsmedel Tvätt- och rengöringsmedel
Concentrated daily cleaner for all water-resistant hard surfaces, such as toilets, tiles and porcelain in washrooms.
  • Concentrated formulation gives excellent economy of use
  • Highly effective removal of common sanitary soil e.g., body fat, cosmetic and soap residues
  • Regular use hinders lime scale and lime soap build-up
  • No fragrance or dye
  • Contains plant-based ingredients from renewable sources
  • Decomposed by natural processes
  • No environmental hazard classifications
SURE® washroom cleaner is part of a unique range of plant-based, cleaning products thatMore Product Information