Floor cleaners, SURE®
Leverantör: Diversey
100891586 100891588
141-1571EA 935 SEK
141-1571 141-1572
Floor cleaners, SURE®
Rengöringsmedel Tvätt- och rengöringsmedel
Concentrated, low foaming daily cleaner for all water resistant hard floors. Recommended for all cleaning methods, including bucket and scrubber drier applications.
  • Concentrated formulation gives excellent economy of use
  • No residues on the floor and no build-up
  • Low foaming effective cleaning
  • No fragrance or dye
  • Contains plant-based ingredients from renewable sources
  • Decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for user
SURE® floor cleaner is part of a unique range of plant-based, cleaning products thatMore Product Information