Tenn 1000 mg/L 20% saltsyra (av Sn) Specpure® standard för ICP

Leverantör: Thermo Fisher Scientific

13863.AD 13863.AE 13863.AP
13863.ADEA 1700 SEK
13863.AD 13863.AE 13863.AP
Tenn 1000 mg/L 20% saltsyra (av Sn) Specpure® standard för ICP
Standarder Cationic Standards Tin Standards

Molecular Formula: Sn in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00133862


Certifikat: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR