Nickel standardlösning, 1000 mg/L Ni in 5% salpetersyra (av Ni) Specpure® standard för ICP

Leverantör: Thermo Fisher Scientific

13839.AD 13839.AE 13839.AP
13839.ADEA 1325 SEK
13839.AD 13839.AE 13839.AP
Nickel standardlösning, 1000 mg/L Ni in 5% salpetersyra (av Ni) Specpure® standard för ICP
Standarder Cationic Standards Nickel Standards

Molecular Formula: Ni in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011137


Certifikat: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR