Industrial label printer, BradyPrinter i3300
Leverantör: Brady
241130 149554 241119 241124 241118 241131 149553
116-0817EA 18950 SEK
116-0817 116-0806 116-0810 116-0813 116-0809 116-0818 116-0805
Industrial label printer, BradyPrinter i3300
Printer Label Printers
Print any label or sign with the versatile BradyPrinter i3300 industrial label printer.
  • Wire and cable identification labels
  • Product and laboratory identification labels
  • Safety and facility signs
The BradyPrinter i3300 Industrial Label Printer is a completely hassle-free, easy to use PC-based printer. It can print a full range of safety and facility labels and signs as well as die-cut labels for wire, product and laboratory identification.
With the BradyPrinter i3300, you can print a huge range of reliable labels and signs at your location inMore Product Information
Order Now
SPECIFIKATIONER
Environmentally Preferable
Display Display colour LCD touchscreen, 272×480 px
Label Continuous tape and die cut labels
Minne 512 MB (RAM), 195 MB (data storage), internal
Upplösning 300 dpi
Mått 240W×317H×435D mm
Vikt 5.30 kg