Aluminiumoxid aktiverad neutral Brockmann Grade I 58 angstroms

Leverantör: Thermo Fisher Scientific

11502.A7 11502.30
11502.A7EA 7500 SEK
11502.A7 11502.30
Aluminiumoxid aktiverad neutral Brockmann Grade I 58 angstroms
Aluminium oxide activated neutral

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: -60 Mesh Powder, S.A. 150m2/g Odour: Odourless Melting Point: 2045° MDL No.: MFCD00003424


Suggested for the separation of certain steroids, alkaloids, hydrocarbons, esters, aldehydes, alcohols and weak organic acids and bases. It is usually used with non-aqueous solutions

Formel: Al₂O₃
MW: 101.96 g/mol
Densitet: 3.94 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003424
CAS nummer: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 14,356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR