Universal snap mounting kit
Leverantör: DESCO
230646
115-3056EA 53 SEK
115-3056
Universal snap mounting kit
Antistatskydd Jordningskabel
The Universal snap mounting kit consists of 10 mm stud and socket, threaded backing washer, conical nut, long and short screw, ESD common ground point label.
Everything needed to field install a stud or socket on all soft materials up to 10 mm. Screws are 6-32 flat head Phillips machine screws; one 6 mm and one 10 mm. Just punch a hole through the mat and screw on the socket or stud, whichever is preferred. All that is needed is a Phillips screwdriver.