Cleanroom wipes, pre-saturated, Polynit Heatseal, Prosat®
Leverantör: CONTEC
PSPS0091 PSPS0047 PSPS0044 PSPS0076-BPR PSPS0077-BPR PSPS0091-BPR
115-2447EA 14525 SEK
115-2447 115-3032 115-2700 115-2817 115-2818 115-2819
Cleanroom wipes, pre-saturated, Polynit Heatseal, Prosat®
Torkdukar Torkdukar, renrum Förindränkt
These heat seal wipes are made from filament polyester that is laser cut to bond the fibres at the edges of the wipes. This creates a very clean, non abrasive edge that allows full utilisation of the wipe surface. The wipes are saturated with 70% USP grade IPA, 30% DI water.
  • Exceptionally low in particles and extractable residue
  • Pre-saturation greatly reduces solvent usage and maximises process control and consistency
  • Abrasion and chemical resistant
Ideal for general purpose and critical cleaning.
Colour: White
Observera: Produkten/produkternaMore Product Information