Cleanroom wipes, sterile, Polynit Heatseal
Leverantör: CONTEC
PN-99IR LWPS0007 LWPS0006 LWPS1004
115-2145EA 14725 SEK
115-2145 115-1862 CONTLWPS0006 CONTLWPS1004
Cleanroom wipes, sterile, Polynit Heatseal
Torkdukar Torkdukar, renrum Steril
100% no-run interlock knit polyester wipes with sealed edges.
  • Validated sterile to a 10⁻⁶ SAL
  • Laundered knitted 100% polyester fabric for extremely low levels of particles and fibres
  • Extremely low levels of particles, fibres and residue
  • Laser-cut, sealed edge prevents release of fibres
  • Laundered and packaged in an ISO Class 4 cleanroom
Sterile Polynit Heatseal wipes are made of 100% knitted polyester that is laser cut to bond the fibres at the edges of the wipe. This creates a very clean non abrasive edge that allows full utilisation of the wipeMore Product Information