Wipes for general applications, Tork

Leverantör: Essity
510104 510137 510237
115-0640EA 3950 SEK
115-0640 115-0338 115-1658
Wipes for general applications, Tork
Torkdukar Torkdukar, återanvändbara Torkduk, universal
Strong and durable cloth wipes for use in a variety of environments, softness and flexibility mean wipes are excellent for cleaning, wiping and polishing even in confined spaces.

  • Cellulose/PP/polyester, reusable
  • Solvent resistant, good dry and wet strength and performance
  • High absorbency
  • Low-linting

Färg: Vit eller blå.

Colour: White or blue
Order Now

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR