Nitrile gloves, Kimtech™ Polaris™ Xtra
Leverantör: KIMBERLY CLARK
62100 62101 62102 62103 62104
112-6565EA 260 SEK
112-6565 112-6566 112-6567 112-6568 112-6569
Nitrile gloves, Kimtech™ Polaris™ Xtra
Handskar
Kimtech™ Polaris™ Xtra Nitrile Gloves are built for science, made and designed for use in laboratories, life science applications, and non-sterile pharmaceutical manufacturing environments.
  • High-end ambidextrous powder-free nitrile gloves deliver seamless protection whenever and wherever it is needed
  • Optimized nitrile formulation (acrylonitrile-butadiene) features extended chemicals and chemotherapy drugs splash protection without compromising comfort
  • Beaded cuffs and textured fingertips enable excellent handling of both wet and dry materials
  • Extensively
More Product Information
Order Now
SPECIFIKATIONER
Environmentally Preferable Waste Reducing
Ej handspecifika/handspecifika Ej handspecifika
Engångs/Återanvändbar Engångs
Latex/latexfri Latexfria
Pudrad/puderfri Puderfria
Yta Textured fingertips
Kragmodell Med rullad kant
Krage, tjocklek 0,11 mil
Finger, tjocklek 0,19 mil
Handflata, tjocklek 0,14 mil
FDA-reglerad +
Draghållfasthet ≥ 14 Mpa
Brottöjning ≥ 400%, min.
EN standard EN ISO 374-1 Type B (JKOPT)
Godkända för livsmedelskontakt +
ASTM nr. D6978