Cleanroom gloves, nitrile, non sterile, light blue, tacky finish, PUREZERO® HG3
Leverantör: Halyard
CLN9031LG CLN9031MD CLN9031SM CLN9031XL CLN9031XS
112-6532EA 5075 SEK
112-6532 112-6533 112-6534 112-6535 112-6536
Cleanroom gloves, nitrile, non sterile, light blue, tacky finish, PUREZERO® HG3
Handskar
High quality gloves designed to exact standards to meet the requirements of a contamination controlled environment.
  • High quality nitrile
  • Non sterile
  • Accelerator-free formula reduces risk of type IV allergies
  • For use in cleanroom environments (ISO 5/6/7/8, grade B/C/D aseptic)
  • REACH Annex XVII
Offering the right glove for the right controlled environment, Halyard PUREZERO® provides a cleanroom glove with guaranteed low particulate counts and endotoxin levels due to repeated washes in deionised water and chlorination. Nickel-, powder-, silicon- andMore Product Information