Pleated masks with earloops, non sterile, HM4, PUREZERO®
Leverantör: Halyard
CLN1341EB
112-6507EA 1750 SEK
112-6507
Pleated masks with earloops, non sterile, HM4, PUREZERO®
Munskydd Munskydd för Renrum
The HM4 Pleated Mask with Earloops is a three-layer mask with nylon earloops that provides a high particle, bacterial, and viral filtration efficiency. The mask construction consists of low-linting materials and a polyester foam nose band.
  • Three-layer mask
  • Nylon earloops
  • Designed for industrial and cleanroom use
  • Recommended for ISO Class 4 and or higher and Grade B/C/D cleanrooms
  • Exclusive SO SOFT biocomponent technology inner lining for comfort
Cleanroom efficiency depends on effective risk management. Ensure masks are up to the task with PUREZERO®More Product Information