VWR® Maximum, Cut Resistant Gloves
Leverantör: VWR Collection
112-5691EA 870 SEK
112-5691 112-5692 112-5693 112-5694 112-5695 112-5696
VWR® Maximum, Cut Resistant Gloves
Handskar
This VWR® glove with polyurethane coating provides superior cut protection for applications that include a high risk of cuts.
  • Nylon/spandex/polyester blend composition
  • Polyurethane (PU) coating on palm and fingertips
  • Grey knitted glove with grey coating
Ideal for handling items or tools with a high risk of cuts, including steel or glass handling, assembly, maintenance, and janitorial applications.
Certifikat: Meets requirements for EN 388:2016 (4X42E). Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in theMore Product Information
Order Now
SPECIFIKATIONER
Environmentally Preferable
Ej handspecifika/handspecifika Handspecifika
Engångs/Återanvändbar Återanvändbar
Latex/latexfri Latexfria
Finish PU palm coating
Mätare 13
Cuff style Stickad mudd
EN standard EN 388:2016 (4X42E)