VWR®, Cleanroom Gloves, Nitrile, Sterile, ISO 5
Leverantör: VWR Collection
112-4549EA 5025 SEK
112-4549 112-4550 112-4551 112-4552 112-4553 112-4554 112-4555
VWR®, Cleanroom Gloves, Nitrile, Sterile, ISO 5
Handskar
These hand-specific nitrile gloves are manufactured from a synthetic copolymer of acrylonitrile, butadiene and methylacrylic acid, that provides reliable, durable and comfortable hand protection during demanding cleanroom applications.
  • Yield exceptionally low particle counts; Grade A; Endotoxin <20 EU/glove
  • Micro textured surface for enhanced grip on components
  • Tapered, beaded cuff provides secure fit
  • Static-dissipative to inhibit harmful static discharge
  • Sterilisation by gamma irradiation to SAL 10⁻⁶
  • Contain no natural rubber latex
  • Silicone-
More Product Information