Coverlite™ Safety spectacle, Stealth™
Leverantör: JSP
ASA940-061-300 ASA940-063-000 ASA940-0A3-000
111-2597EA 120 SEK
111-2597 111-2598 111-2599
Coverlite™ Safety spectacle, Stealth™
Skyddsglasögon
Stylish lightweight overspec for use with prescription eyewear.
  • Stylish overspec
  • Optically perfect
  • Pantoscopic soft grip arms
  • Flexible frame
  • Ultra lightweight
The stylish and spacious design of the Stealth™ Coverlite™ allows them to comfortably fit over prescription eyewear while maintaining a modern look, unlike traditional bulkier overspectacles. The frame is flexible and springy so that they can fit any head size and create a secure fit. The robust, low base, minimum curve, optical Class 1 lens provides optimal vision to the wearer.