Respirator filters, TR-300+ Series, for Versaflo™ powered air respirator
Leverantör: 3M
TR-3822E TR-3802E TR3712E
MMMATR-3822EEA 4300 SEK
MMMATR-3822E 111-2403 111-2409
Respirator filters, TR-300+ Series, for Versaflo™ powered air respirator
Masker Andningsskydd, återanvändbara Maskfilter
These filters provide protection against dusts, mists and other particulate hazards when used with the 3M™ Versaflo Powered Air Respirator, TR-300 Series, and a 3M™ approved headtop.
  • Filters can be exchanged quickly and easily
  • Filters need replacing at required filter interval decided by risk assesment
TR-3712E - Particulate (P) filter for protection against dusts, mists, fumes and other particulate hazards.
TR-3802E - Particulate (P) filter for protection against dusts, mists, fumes and other particulate hazards + nuisance level Organic Vapours (More Product Information