Natriumbromid 99,9955 Metaller basis, Puratronic®

Leverantör: Thermo Fisher Scientific
Varning

10860.14 10860.22
10860.14EA 3450 SEK
10860.14 10860.22
Natriumbromid 99,9955 Metaller basis, Puratronic®
Sodium bromide

Purity: 99.9955% CAS No.: 7647-15-6 Molecular Formula: NaBr Formula Weight: 102.89 Physical Form: Powder Density: 3.203 Refractive Index: 1.6412 Sensitivity: Hygroscopic Melting Point: 755° Boiling Point: 1390° Merck Reference: 14,8594 MDL No.: MFCD00003475

Formel: NaBr
MW: 102.89 g/mol
Kokpunkt: 1390 °C
Smältpunkt: 755 °C
Densitet: 3.203
Storage Temperature: Omgivande
MDL Number: MFCD00003475
CAS nummer: 7647-15-6
EINECS: 231-599-9
Merck Index: 14,8594

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR