β-glucuronidase substrates
Leverantör: Biotium
10020 10018 10019 10017 10025
10020.EA 2100 SEK
10020. 10018. 10019. 10017. 10025.
β-glucuronidase substrates
Infärgningsmedel Färgämnen för in vivo bildbehandling
β-glucuronidase substrates are easy to detect stains against the background of plant cells.
White solid soluble in water or DMSO.
Observera: Store at −20 °C and protect from light.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34e0c5631768404e7b136dcd4c8ccea5ff193363&vid=34e0c5631768404e7b136dcd4c8ccea5ff193363&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ed27d935525d00de7934081ea675472f98700b5&vid=2ed27d935525d00de7934081ea675472f98700b5&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff392c05de91ded87a0519a5368c79427c99dc88&vid=ff392c05de91ded87a0519a5368c79427c99dc88&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f3ae967dad2e8db89a4809f5c4b6129802575e6&vid=6f3ae967dad2e8db89a4809f5c4b6129802575e6&prid=web&clid=SAPDAM