Patent Blue VF (Disulphine blue), Supelco®

Leverantör: Merck

Synonyms: acid Blue1, Patent Blue, Patent Blue VF, Hydrogen [4-[4-(diethylamino)-2',4'-disulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene]diethylammonium, sodium salt, Patent blue VF sodium salt

MQ200

1.12144.0025
1.12144.0025EA 1380 SEK
1.12144.0025
Patent Blue VF (Disulphine blue), Supelco®
Patent Blue VF (Disulphine blue)
Formel: C₂₇H₃₁N₂NaO₆S₂
MW: 566.67 g/mol
Smältpunkt: 290
Storage Temperature: Omgivande
MDL Number: MFCD00012119
CAS nummer: 129-17-9
EINECS: 204-934-1
Merck Index: 12,09161

Order Now

Specifikation testresultat

Solubility 210 g/l (25 °C)
Melting Point 290 °C (decomposition)
Molar Mass 566.67 g/mol
Bulk Density 600 kg/m3
pH 6.9 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR