Saltsyra 5 mol/l (5 N), Titrisol®, Supelco®

Leverantör: Merck
Varning

Synonyms: Hydrochloric acid solution, Muriatic acid

M-Clarity™ quality level = MQ100

1.09911.0001
1.09911.0001EA 489.75 SEK
1.09911.0001
Saltsyra 5 mol/l (5 N), Titrisol®, Supelco®
Saltsyra
Formel: HCl
MW: 36.46 g/mol
Densitet: 1.06 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8

Order Now

Specifikation testresultat

Solubility (20 °C) soluble
Density 1.1 g/cm3 (20 °C)
pH  - 1 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR