2-Metyl-2-propanol, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®

Leverantör: Merck
Fara

Synonyms: tert-Butylalkohol

MQ300

1.09629.0500 1.09629.9025
1.09629.0500EA 784 SEK
1.09629.0500 1.09629.9025
2-Metyl-2-propanol, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®
2-Metyl-2-propanol
Formel: (CH₃)₃COH
MW: 74.12 g/mol
Kokpunkt: 82.2
Smältpunkt: 25.3
Densitet: 0.7887 g/cm³ (20 °C)
Flampunkt: 11
Storage Temperature: Omgivande
MDL Number: MFCD00004464
CAS nummer: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3 II
Merck Index: 12,01577
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a1b0c51eec643e5d5b7152a3e410db93d1626e&vid=999328a3c31895b190d09313b24501682921af01&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Solubility (20 °C) soluble
Melting Point 24 - 25 °C
Molar Mass 74.12 g/mol
Boiling Point 81 - 83 °C (1013 hPa)
Vapor Pressure 40.7 hPa (20 °C)
Flash Point 14 °C
Explosion Limit 2.3 - 8.0 %(V)
Density 0.78 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 490 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR