Benzylalkohol, EMSURE® analysis_for, Supelco®

Leverantör: Merck
Varning

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

MQ300

1.09626.1000 1.09626.2500 1.09626.6025
1.09626.1000EA 2330 SEK
1.09626.1000 1.09626.2500 1.09626.6025
Benzylalkohol, EMSURE® analysis_for, Supelco®
Benzylalkohol
Formel: C₆H₅CH₂OH
MW: 108.14 g/mol
Kokpunkt: 205
Smältpunkt: –15.3 °C
Densitet: 1.05 g/cm³ (20 °C)
Flampunkt: 101
Storage Temperature: Omgivande
MDL Number: MFCD00004599
CAS nummer: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Solubility 40 g/l (20 °C)
Melting Point -15.3 °C
Molar Mass 108.14 g/mol
Boiling Point 205 °C (1013 hPa)
Vapor Pressure 0.03 hPa (20 °C)
Flash Point 101 °C DIN 51758
Refractive Index 1.54
Explosion Limit 1.3 - 13 %(V)
Density 1.05 g/cm3 (20 °C)
Ignition Point 435 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR