Natriumhydroxid 0.25 mol/l (0.25 N) aqueous solution, Titripur® Volumetric, Supelco®

Leverantör: Merck
Varning

Synonyms: Natronlut, Kaustik soda

MQ100

1.09139.1000 1.09139.9010 1.09139.1003
1.09139.1000EA 347 SEK
1.09139.1000 1.09139.9010 1.09139.1003
Natriumhydroxid 0.25 mol/l (0.25 N) aqueous solution, Titripur® Volumetric, Supelco®
Natriumhydroxid

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: NaOH
MW: 40 g/mol
Kokpunkt: ∼105 °C (1013 hPa)
Smältpunkt: 323
Densitet: 1.02 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003548
CAS nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specifikation testresultat

Form liquid
Amount-of-substance concentration 0.24875 - 0.25125
Measurement uncertainty 0.00075
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR