Toluen, LiChrosolv® för HPLC, Supelco®

Leverantör: Merck
Fara

Synonyms: Methylbenzene, Phenylmethane, FM approval

M-Clarity™ quality level = MQ100

1.08327.1000 1.08327.4000 1.08327.2500
1.08327.1000EA 799 SEK
1.08327.1000 1.08327.4000 1.08327.2500
Toluen, LiChrosolv® för HPLC, Supelco®
Toluen
Formel: C₆H₅CH₃
MW: 92.14 g/mol
Kokpunkt: 110,6
Smältpunkt: –95 °C
Densitet: 0.867 g/cm³ (20 °C)
Flampunkt: 4
Storage Temperature: Omgivande
MDL Number: MFCD00008512
CAS nummer: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Limited Time Sale

Select items below are currently on sale

Order Now

Specifikation testresultat

Solubility 0.52 g/l (20 °C)
Melting Point -95 °C
Molar Mass 92.14 g/mol
Boiling Point 110.6 °C (1013 hPa)
Vapor Pressure 29 hPa (20 °C)
Flash Point 4 °C
Refractive Index 1.4968 (20 °C)
Explosion Limit 1.2 - 8 %(V)
Density 0.87 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 535 °C
Viscosity kinematic 0.7 mm2/s (20 °C)
Synonyms Toluol, Methylbenzene
Hill Formula C7H8
Chemical Formula C6H5CH3

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR