Sputofluol® Microbiology, Microscopy, Sigma-Aldrich®

Leverantör: Merck
Fara


MQ300

1.08000.1000
1.08000.1000EA 886 SEK
1.08000.1000
Sputofluol® Microbiology, Microscopy, Sigma-Aldrich®
Sputofluol®

Sputofluol® can be used as pre-treatment solution for microscopy investigation of mycobacteria. It will release the mycobacteria from sputum and other material of human origin through the oxidative dissolution of organic material.


It is also part of the Tb-color kit and can be used in clinical diagnostics as it is CE certified and registered as IVD.

Storage Temperature: Omgivande
UN: 1791
ADR: 8 II
IVDLogo

Order Now

Specifikation testresultat

NaOH content (acidimetric) 7 - 8%
Assay (iodometric, free Cl₂) ≥4,5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR