Natriumnitrat 99,99 Metaller basis, Suprapur® Trace_analysis, Supelco®

Leverantör: Merck
Varning


MQ100

1.06546.0050
1.06546.0050EA 3130 SEK
1.06546.0050
Natriumnitrat 99,99 Metaller basis, Suprapur® Trace_analysis, Supelco®
Sodium nitrate
Formel: NaNO₃
MW: 84.99 g/mol
Kokpunkt: 380
Smältpunkt: 308
Densitet: 2.26 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011119
CAS nummer: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1 III
REACH: 01-2119488221-41
Merck Index: 13,08720

Order Now

Specifikation testresultat

Solubility 874 g/l (20 °C)
Melting Point 308 °C
Molar Mass 84.99 g/mol
Bulk Density 1200 kg/m3
Density 2.26 g/cm3 (20 °C)
pH 5.5 - 8.0 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR