2,6-Dichlorophenolindophenol sodium salt dihydrate 98 Torr_basis, GR analysis_for, Supelco®

Leverantör: Merck

Synonyms: 2,6-Dichloroindophenol, sodium salt dihydrate, Sodium 4-(3,5-dichloro-4-oxocyclohexa-2,5-dienylideneamino)phenoxide dihydrate, Tillmans' reagent dihydrate, 2,6-Dichloroindophenol sodium dihydrate, 2,6-Dichlorophenolindophenol sodium salt, 2,6-Dichloroindophenol sodium salt dihydrate, Sodium 2,6-Dichlorobenzenone-indo-phenol

MQ100

1.03028.0005 1.03028.0025
1.03028.0005EA 2770 SEK
1.03028.0005 1.03028.0025
2,6-Dichlorophenolindophenol sodium salt dihydrate 98 Torr_basis, GR analysis_for, Supelco®
2,6-Dichlorophenolindophenol sodium salt dihydrate

For analysis for the determination of ascorbic acid

Formel: C₁₂H₆Cl₂NNaO₂·2H₂O
MW: 326.105 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00150015
CAS nummer: 620-45-1
EINECS: 210-640-4

Order Now

Specifikation testresultat

Solubility (20 °C) slightly soluble
Molar Mass 326.11 g/mol
Bulk Density 320 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR