N-Metyl-2-pyrrolidon (NMP), SupraSolv® GC_HS

Leverantör: Merck
Fara

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone

MQ100

1.02497.0500 1.02497.1000 1.02497.2500
1.02497.0500EA 1600 SEK
1.02497.0500 1.02497.1000 1.02497.2500
N-Metyl-2-pyrrolidon (NMP), SupraSolv® GC_HS
N-Metyl-2-pyrrolidon (NMP)
Formel: C₅H₉NO
MW: 99.13 g/mol
Kokpunkt: 202
Smältpunkt: –24 °C
Densitet: 1.033 g/cm³ (20 °C)
Flampunkt: 91
Storage Temperature: Omgivande
MDL Number: MFCD00003193
CAS nummer: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR