Krom(III)klorid hexahydrat

Leverantör: Merck
Varning

Synonyms: Chromium trichloride hexahydrate, Chromic chloride hexahydrate

1.02487.1000 1.02487.9050
1.02487.1000EA 1825 SEK
1.02487.1000 1.02487.9050
Krom(III)klorid hexahydrat
Krom(III)klorid hexahydrat
Formel: CrCl₃·6H₂O
MW: 266.45 g/mol
Smältpunkt: 85
Densitet: 1.76 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149660
CAS nummer: 10060-12-5
EINECS: 233-038-3
UN: 3260
ADR: 8 III
Merck Index: 13,02242

Order Now

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR