Bariumklorid dihydrat, Supelco®

Leverantör: Merck
Fara

Synonyms: Barium dichloride

MQ200

1.01717.1000 1.01717.9050
1.01717.1000EA 1140 SEK
1.01717.1000 1.01717.9050
Bariumklorid dihydrat, Supelco®
Bariumklorid dihydrat
Formel: BaCl₂·2H₂O
MW: 244.27 g/mol
Kokpunkt: 1560
Smältpunkt: 960
Densitet: 3.86 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149154
CAS nummer: 10326-27-9
EINECS: 233-788-1
UN: 1564
ADR: 6.1 III
Merck Index: 14,00971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR