Silversulfat 80 g/l sulphuric acid COD_determination, Supelco®

Leverantör: Merck
Fara

Synonyms: Disilver sulphate

MQ100

1.01574.1000
1.01574.1000EA 5370 SEK
1.01574.1000
Silversulfat 80 g/l sulphuric acid COD_determination, Supelco®
Silversulfat

For COD determination acc. to DIN 38409-H 43-1

Formel: Ag₂SO₄
MW: 311.8 g/mol
Kokpunkt: 1085
Smältpunkt: 655
Storage Temperature: Omgivande
MDL Number: MFCD00003407
CAS nummer: 10294-26-5
EINECS: 233-653-7
UN: 3264
ADR: 8 II
Merck Index: 13,08603
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Concentration (Ag₂SO₄) 78.8 - 81.2 g/l
Concentration (H₂SO₄) 1665 - 1735 g/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR