Metylorange 0,1% i vattenlösning, Supelco®

Leverantör: Merck

Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine

M-Clarity™ quality level = MQ200

1.01323.0250 1.01323.1000
1.01323.0250EA 1040 SEK
1.01323.0250 1.01323.1000
Metylorange 0,1% i vattenlösning, Supelco®
Metylorange
Formel: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Kokpunkt: 90
Flampunkt: 38
Storage Temperature: Omgivande
MDL Number: MFCD00007502
CAS nummer: 547-58-0
EINECS: 208-925-3
Merck Index: 13,06127

Order Now

Specifikation testresultat

Solubility (20 °C) soluble
Density 1.00 g/cm3 (20 °C)
pH 6 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR