Dietyleter, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®

Leverantör: MERCK PRODUCTION CHEMICALS
Fara

Synonyms: Eter

Stabilization: 5 - 10 ppm Stabilised med BHT/Ionol
M-Clarity™ quality level = MQ500

1.00926.1000 1.00926.6025
1.00926.1000EA 1750 SEK
1.00926.1000 1.00926.6025
Dietyleter, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Dietyleter
Formel: (CH₃CH₂)₂O
MW: 74.12 g/mol
Kokpunkt: 35
Smältpunkt: –123 °C
Densitet: 0.7135 g/cm³ (20 °C)
Flampunkt: –40 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011646
CAS nummer: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3 I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (GC) ≥ 99.5 %
Identity (IR) conforms
Acidic substances conforms
Density (d 20 °C/20 °C) 0.714 - 0.716
Boiling point (lower value) ≥ 34.0 °C
Boiling point (upper value) ≤ 35.0 °C
Foreign odor conforms
Aldehydes conforms
Peroxides conforms
Residual solvents (ICH Q3C) excluded by production process
Evaporation residue ≤ 20 mg/l
Water ≤ 0.20 %
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities).
Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Stabilized with 5 - 10 ppm 2,6-Di-tert-butyl-4-methylphenol (BHT).
Conforms to Ph Eur, BP

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR