Benzaldehyd, Supelco®

Leverantör: Merck
Varning


MQ100

09143-25ML-F 09143-5ML-F
09143-25ML-FEA 3250 SEK
09143-25ML-F 09143-5ML-F
Benzaldehyd, Supelco®
Benzaldehyd
Formel: C₆H₅CHO
MW: 106.12 g/mol
Kokpunkt: 179
Smältpunkt: –26 °C
Densitet: 1.049 g/cm³ (20 °C)
Flampunkt: 64
Storage Temperature: Omgivande
MDL Number: MFCD00003299
CAS nummer: 100-52-7
EINECS: 202-860-4
UN: 1990
ADR: 9 III
Merck Index: 13,01057
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6554d2d6d77311eee8e84bfe276729e9af4346b&vid=daf04a77c3527e1c8a257ef2d911d69c6f6ccc10&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR