Germanium 99,9999 Metaller basis, pieces ≤ 2 cm (0.8 in)

Leverantör: Thermo Fisher Scientific

00123.09 00123.18
00123.09EA 4425 SEK
00123.09 00123.18
Germanium 99,9999 Metaller basis, pieces ≤ 2 cm (0.8 in)
Germanium

Purity: 99.9999% CAS No.: 7440-56-4 Physical Form: Electrical Resistivity: 40 ohm-cm Odour: Odourless MDL No.: MFCD00085310

Formel: Ge
MW: 72.61 g/mol
Kokpunkt: 2830
Smältpunkt: 937.2
Densitet: 5.323 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00085310
CAS nummer: 7440-56-4
EINECS: 231-164-3
Merck Index: 12,04415

Order Now

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR