2-Bromoterephthalic acid ≥98.0%

Leverantör: TCI
Varning

B1321-25G B1321-5G
TCIAB1321-25GEA 6775 SEK
TCIAB1321-25G TCIAB1321-5G
2-Bromoterephthalic acid ≥98.0%
2-Bromoterephthalic acid
Formel: C₈H₅BrO₄
MW: 245,03 g/mol
MDL Number: MFCD00002403
CAS nummer: 586-35-6
EINECS: 209-572-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b56ad4dece402518f369ecddbe26c84defaf93c1&vid=b56ad4dece402518f369ecddbe26c84defaf93c1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR