1-Decyne ≥95.0%

Leverantör: TCI
Fara

D0037-25ML D0037-5ML
TCIAD0037-25MLEA 765 SEK
TCIAD0037-25ML TCIAD0037-5ML
1-Decyne ≥95.0%
1-Decyne
Formel: C₁₀H₁₈
Kokpunkt: 174°C
Smältpunkt: –44 °C
Densitet: 0,766 g/cm³ (20 °C)
Flampunkt: 50°C
Storage Temperature: Kylskåp
MDL Number: MFCD00009576
CAS nummer: 764-93-2
EINECS: 212-132-8
UN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c53acc93d8422f63145fe4e41115bc66cc94e4ae&vid=26140693591997a48877302e73e973230f982c1b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR