Dropping bottles, LDPE, Azlon®
Leverantör: SciLabware
BGE514P BGE520P BGE528P BGE530P BGE536P
215-0472EA 385 SEK
215-0472 215-0473 215-0474 215-0475 215-0476
Dropping bottles, LDPE, Azlon®
Flaskor Droppflaskor
LDPE, translucent.
  • Good chemical resistance
  • Heat resistant up to +80 °C
  • Easy handling
Nozzle retracts into the cap to seal the bottle. Suitable for use as a dropping or wash bottle and excellent for dispensing gels.