4-Nitrophenylarsonic acid ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI
Fara

Synonyms: Nitarsone, 4-Nitrobenzenearsonic acid, 4-Nitrophenylarsonic acid

N0964-200MG N0964-1G
TCIAN0964-200MGEA 1425 SEK
TCIAN0964-200MG TCIAN0964-1G
4-Nitrophenylarsonic acid ≥98.0% (by HPLC, titration analysis)
4-Nitrophenylarsonic acid
Formel: C₆H₆AsNO₅
MW: 247,04 g/mol
Smältpunkt: 300°C
MDL Number: MFCD00024575
CAS nummer: 98-72-6
EINECS: 202-695-8
UN: 3465
ADR: 6.1,III
Merck Index: 14,06566
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1969df527c0446df4dce652bc477273e60d434c2&vid=1969df527c0446df4dce652bc477273e60d434c2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR