NU 2058 ≥98.0%

Leverantör: TCI
Varning

Synonyms: NU2058, O-Cyclohexylmethylguanine, 6-(Cyclohexylmethoxy)-9H-purin-2-amine

N0958-20MG N0958-100MG
TCIAN0958-20MGEA 615 SEK
TCIAN0958-20MG TCIAN0958-100MG
NU 2058 ≥98.0%
NU 2058
Formel: C₁₂H₁₇N₅O
MW: 247,30 g/mol
Smältpunkt: 199°C
Storage Temperature: Kylskåp
MDL Number: MFCD05664734
CAS nummer: 161058-83-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05aeb54196469f3e242ce738f9ba8025ff3c9478&vid=05aeb54196469f3e242ce738f9ba8025ff3c9478&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 27.00 to 29.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR